Learn More
Medchemexpress LLC IL-11R alpha Mouse 10ug
SDP
Catalog No. 5000196824
Encompass_Preferred
Recombinant mouse interleukin-11 receptor subunit alpha (IL-11Rα), expressed in HEK293 cells with a polyhistidine tag, provided as a research reagent for studies of IL-11 signaling, receptor binding, and biochemical assays. The sequence corresponds to amino acids S24-Q367 (accession Q64385). Endotoxin is controlled and storage and reconstitution guidance are provided with the product.
  • Recombinant mammalian expression for native-like post-translational modifications.
  • Includes a C-terminal His tag to facilitate purification and detection.
  • Covers amino acids S24-Q367 corresponding to the extracellular domain.
  • Low endotoxin level suitable for cell-based assays.
  • Approximate molecular weight 40-55 kDa by SDS-PAGE.
  • Stable at -20°C for long-term storage; reconstitution guidance provided.
Catalog No. 50-001-96824 Supplier Medchemexpress LLC Supplier No. HYP7086010UG
Recombinant mouse interleukin-11 receptor subunit alpha (IL-11Rα), expressed in HEK293 cells with a polyhistidine tag, provided as a research reagent for studies of IL-11 signaling, receptor binding, and biochemical assays. The sequence corresponds to amino acids S24-Q367 (accession Q64385). Endotoxin is controlled and storage and reconstitution guidance are provided with the product.
  • Recombinant mammalian expression for native-like post-translational modifications.
  • Includes a C-terminal His tag to facilitate purification and detection.
  • Covers amino acids S24-Q367 corresponding to the extracellular domain.
  • Low endotoxin level suitable for cell-based assays.
  • Approximate molecular weight 40-55 kDa by SDS-PAGE.
  • Stable at -20°C for long-term storage; reconstitution guidance provided.