Learn More
Medchemexpress LLC IL-12 beta Marmoset 2ug | 2ug
SDP
Catalog No. 5000187145
Encompass_Preferred
IL-12 beta Protein also known as natural killer cell stimulatory factor 2 is a common subunit (p40) of IL-12 and IL-23 IL-12 is a inflammatory factor expressed by activated macrophages and involves in Th1-type immune response against cancer[1] IL-12 beta Protein located outside the cell membrane involves in singalling mediated by Jak-STAT[3] IL-12 beta Protein consists of 335 amino acids (M1-N328) with a fibronectin type-III domain (233-324 a a) IL-12 beta Protein Marmoset is produced in HEK293 cells with a C-Terminal hFc-tag
Catalog No. 50-001-87145 Supplier Medchemexpress LLC Supplier No. HYP774022UG
IL-12 beta Protein also known as natural killer cell stimulatory factor 2 is a common subunit (p40) of IL-12 and IL-23 IL-12 is a inflammatory factor expressed by activated macrophages and involves in Th1-type immune response against cancer[1] IL-12 beta Protein located outside the cell membrane involves in singalling mediated by Jak-STAT[3] IL-12 beta Protein consists of 335 amino acids (M1-N328) with a fibronectin type-III domain (233-324 a a) IL-12 beta Protein Marmoset is produced in HEK293 cells with a C-Terminal hFc-tag