Learn More
Medchemexpress LLC HY-P7208 50ug Medchemexpress, IL-12 (CHO-expressed), Mouse CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP720850UG
Encompass_Preferred
Medchemexpress, HY-P7208 50ug IL-12 (CHO-expressed), Mouse CAS: Recombinant Mouse Interleukin-12 (CHO-expressed) has both T cell growth and IFN-γ stimulatory properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4363