Learn More
Medchemexpress LLC IL-12 Human Biotin 20ug | 20ug
SDP
Catalog No. 5000187079
Encompass_Preferred
IL-35 protein plays a key role in immune regulation forming IL-12 cytokine with IL12B or IL-35 cytokine with EBI3/IL27B IL-12 modulates T cell and natural killer cell responses and induces interferon gamma production IL-12 Protein Human (Biotinylated HEK293 His-Avi) is a recombinant protein dimer complex containing human-derived IL-12 protein expressed by HEK293 with C-Avi C-His labeled tag IL-12 Protein Human (Biotinylated HEK293 His-Avi) has molecular weight of 40-45 kDa
Catalog No. 50-001-87079 Supplier Medchemexpress LLC Supplier No. HYP7770220UG
IL-35 protein plays a key role in immune regulation forming IL-12 cytokine with IL12B or IL-35 cytokine with EBI3/IL27B IL-12 modulates T cell and natural killer cell responses and induces interferon gamma production IL-12 Protein Human (Biotinylated HEK293 His-Avi) is a recombinant protein dimer complex containing human-derived IL-12 protein expressed by HEK293 with C-Avi C-His labeled tag IL-12 Protein Human (Biotinylated HEK293 His-Avi) has molecular weight of 40-45 kDa