Learn More
Medchemexpress LLC IL-12 Human HEK293 100ug
SDP
Catalog No. 5000179790
Encompass_Preferred
Interleukin-12 subunit alpha (IL-12A IL-12p35) an immune-suppressive cytokine encodes a subunit of the cytokine IL-12 that acts on T and natural killer cells and has a broad array of biological activities IL-12A heterodimerizes with IL-12B to form the IL-12 cytokine or with EBI3/IL27B to form the IL-35 cytokine[1][2] IL-12 Protein Human (547a a HEK293 His) is produced in HEK293 cells with a C-Terminal His-tag It consists of IL-12A (M1-S219) and IL-12B (M1-S328)
Catalog No. 50-001-79790 Supplier Medchemexpress LLC Supplier No. HYP73898100UG
Interleukin-12 subunit alpha (IL-12A IL-12p35) an immune-suppressive cytokine encodes a subunit of the cytokine IL-12 that acts on T and natural killer cells and has a broad array of biological activities IL-12A heterodimerizes with IL-12B to form the IL-12 cytokine or with EBI3/IL27B to form the IL-35 cytokine[1][2] IL-12 Protein Human (547a a HEK293 His) is produced in HEK293 cells with a C-Terminal His-tag It consists of IL-12A (M1-S219) and IL-12B (M1-S328)