Learn More
Medchemexpress LLC IL-12 protein, human (HEK293) | >95.0% | 100UG
SDP
Catalog No. 5000174467
Encompass_Preferred
IL-12 protein (human, HEK293) is a recombinant, heterodimeric pro-inflammatory cytokine produced in HEK293 cells. It comprises the p35 and p40 subunits, is supplied tag-free and lyophilized, and is intended for research into cell-mediated immunity, induction of IFNγ, activation of T and NK cells, and TH1 differentiation. Refer to the lot-specific COA for exact buffer and storage information.
  • Purity >95% as determined by reducing SDS-PAGE.
  • Expression in HEK293 preserves human post-translational modifications.
  • Includes both p35 and p40 subunits for functional activity.
  • Supplied lyophilized for convenient storage and transport.
  • Stable when aliquoted and stored at -20°C or -80°C for extended periods.
Catalog No. 50-001-74467 Supplier Medchemexpress LLC Supplier No. HYP7032100UG
IL-12 protein (human, HEK293) is a recombinant, heterodimeric pro-inflammatory cytokine produced in HEK293 cells. It comprises the p35 and p40 subunits, is supplied tag-free and lyophilized, and is intended for research into cell-mediated immunity, induction of IFNγ, activation of T and NK cells, and TH1 differentiation. Refer to the lot-specific COA for exact buffer and storage information.
  • Purity >95% as determined by reducing SDS-PAGE.
  • Expression in HEK293 preserves human post-translational modifications.
  • Includes both p35 and p40 subunits for functional activity.
  • Supplied lyophilized for convenient storage and transport.
  • Stable when aliquoted and stored at -20°C or -80°C for extended periods.