Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Porcine IL-12 (IL-12A) ELISA Kit

Catalog No. ESIL12A
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ESIL12A 96 Tests
50-254-5274 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ESIL12A Supplier Invitrogen™ Supplier No. ESIL12A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Porcine Interleukin 12 (IL-12A) ELISA quantitates Porc IL-12A in porcine serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Porc IL-12A. Principle of the method The Porcine IL-12A solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Interleukin 12A is a novel signaling cytokine belonging to the Interleukin-6 protein superfamily. Interleukin-12 alpha chain precursor (IL-12A) is a 35-kD subunit of disulfide-linked heterodimer of cytokine IL-12 and has a broad array of biological activities. Reports suggest that IL-12A can act as a growth factor for activated T and NK cells and enhance the lytic activity of NK/lymphokine-activated Killer cells. IL-12A stimulates the production of IFN-gamma by resting PBMC as well as T-cell-independent induction of interferon (IFN)-gamma, and helps in the differentiation of both Th1 and Th2 cells. It is also involved in positive regulation of tyrosine phosphorylation of Stat4 protein. The signaling process of IL-12A in innate immunity requires nitric oxide synthase 2A. Along with EBI3, IL-12A forms a novel heterodimeric cytokine, designated as interleukin-35 (IL-35), that may be specifically produced by T reg cells and is required for maximal suppressive activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q29053
Assay Range 2.2-600 ng/mL
Assay Sensitivity 2.2 ng/mL
Calibration Range 2.2 ng/ml to 600 ng/ml
Content And Storage Pre-coated 96-well plate, standard, detector antibody, streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers. May be stored at 2–8°C for up to 6 months from the date of shipment.
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Cytokines & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: porcine IL-1 beta, IL-4, IL-6, IL-8, IL-10, IL-13, GM-CSF, IFN-gamma, TGF beta 1, TNF alpha
Sufficient For 96 assays
Target Species Pig
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias CLMF; CLMF p35; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; il 12; IL-12 subunit p35; IL-12, subunit p35; Il12a; IL-12A; IL12p35; IL-12p35; IL35 subunit; ILN; Interleukin; Interleukin 12; interleukin 12 p35 subunit; interleukin 12, p35; interleukin 12a; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); Interleukin12; interleukin-12 alpha chain; interleukin-12 beta p35 subunit; interleukin-12 p35 subunit; interleukin-12 subunit alpha; Ll12a; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NKSF1; P35CLMF; CLMF p35; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; il 12; IL-12 subunit p35; IL-12, subunit p35; Il12a; IL-12A; IL12p35; IL-12p35; IL35 subunit; ILN; Interleukin; Interleukin 12; interleukin 12 p35 subunit; interleukin 12, p35; interleukin 12a; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); Interleukin12; interleukin-12 alpha chain; interleukin-12 beta p35 subunit; interleukin-12 p35 subunit; interleukin-12 subunit alpha; Ll12a; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NKSF1; P35;
Gene ID (Entrez) 397053
Gene Symbol IL12A
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases IL-12A, Cytotoxic lymphocyte maturation factor 35 kDa subunit, CLMF p35, IL-12 subunit p35
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Oncology
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target IL-12 p35
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.