Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-12 Mouse CHO 10ug
SDP

Catalog No. 5000195537
Encompass_Preferred

Recombinant mouse interleukin-12 (IL-12) produced in CHO cells; a biologically active heterodimeric cytokine used to stimulate T cell proliferation and interferon-gamma production in immunology research. Supplied as a lyophilized powder in small research pack sizes for functional assays, ELISA, and cell culture studies.

  • Recombinant IL-12 expressed in CHO cells.
  • Heterodimeric cytokine with T cell growth and IFN-γ stimulatory activity.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized powder format for long-term storage and easy reconstitution.
  • Available in small research pack sizes suitable for assay development.
  • Stable at -20°C for long-term storage; aliquot after reconstitution.

Catalog No. 50-001-95537 Supplier Medchemexpress LLC Supplier No. HYP720810UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse interleukin-12 (IL-12) produced in CHO cells; a biologically active heterodimeric cytokine used to stimulate T cell proliferation and interferon-gamma production in immunology research. Supplied as a lyophilized powder in small research pack sizes for functional assays, ELISA, and cell culture studies.

  • Recombinant IL-12 expressed in CHO cells.
  • Heterodimeric cytokine with T cell growth and IFN-γ stimulatory activity.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized powder format for long-term storage and easy reconstitution.
  • Available in small research pack sizes suitable for assay development.
  • Stable at -20°C for long-term storage; aliquot after reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.