Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-12 Mouse HEK293 100ug
SDP

Catalog No. 5000184546
Encompass_Preferred

Recombinant mouse interleukin-12 (IL-12) produced in HEK293 mammalian cells and supplied tag-free for research use. The heterodimeric cytokine is provided lyophilized with defined buffer formulations and high purity, suitable for cell activation assays, cytokine signaling studies, and other immunology research applications.

  • Produced in HEK293 mammalian expression system.
  • Tag-free heterodimeric cytokine.
  • Available in multiple sizes, including 100 μg.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized formulation for stability and ease of storage.
  • Recommended aliquoting and storage at -20°C or -80°C for extended preservation.

Catalog No. 50-001-84546 Supplier Medchemexpress LLC Supplier No. HYP70666100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse interleukin-12 (IL-12) produced in HEK293 mammalian cells and supplied tag-free for research use. The heterodimeric cytokine is provided lyophilized with defined buffer formulations and high purity, suitable for cell activation assays, cytokine signaling studies, and other immunology research applications.

  • Produced in HEK293 mammalian expression system.
  • Tag-free heterodimeric cytokine.
  • Available in multiple sizes, including 100 μg.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized formulation for stability and ease of storage.
  • Recommended aliquoting and storage at -20°C or -80°C for extended preservation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.