Learn More
Medchemexpress LLC IL-12 Mouse HEK293 2ug
SDP
Catalog No. 5000184433
Encompass_Preferred
IL-12 Protein, Mouse (HEK293) is a recombinant heterodimeric cytokine produced in HEK293 mammalian cells and supplied lyophilized in microgram-scale sizes. The tag-free protein is >95% pure by reducing SDS-PAGE and is provided with sequence, formulation, and storage guidance for use in cell culture, functional assays, and immunology research.
  • Produced in HEK293 mammalian expression system for native post-translational modifications.
  • Tag free to minimize assay interference.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Lyophilized formulation for long-term stability and easy storage.
  • Includes recommended storage and reconstitution guidance to preserve activity.
  • Available in microgram sizes suitable for assay development and pilot studies.
Catalog No. 50-001-84433 Supplier Medchemexpress LLC Supplier No. HYP706662UG
IL-12 Protein, Mouse (HEK293) is a recombinant heterodimeric cytokine produced in HEK293 mammalian cells and supplied lyophilized in microgram-scale sizes. The tag-free protein is >95% pure by reducing SDS-PAGE and is provided with sequence, formulation, and storage guidance for use in cell culture, functional assays, and immunology research.
  • Produced in HEK293 mammalian expression system for native post-translational modifications.
  • Tag free to minimize assay interference.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Lyophilized formulation for long-term stability and easy storage.
  • Includes recommended storage and reconstitution guidance to preserve activity.
  • Available in microgram sizes suitable for assay development and pilot studies.