Learn More
AdipoGen IL-12 msFc-KIH h rec
SDP
Catalog No. 502562820
Encompass_Preferred
Recombinant protein. IL-12A/p35 aa 23-215 mouseFc Knobs and IL-12B/p40 aa 23-335 mouseFc Holes form the IL-12 mouseFc-KIH human using the Knobs-into-Holes technology see reference J.B. Ridgway, et al. Protein Eng. 9, 617 1996. Source CHO cells. Lyophilized. Contains PBS Interleukin-12 IL-12 family members are heterodimer glycoproteins, composed of two covalently linked subunits, alpha and beta chains. The alpha-subunit consists of IL-23p19, IL-27p28 and IL-12p35, and the beta-subunit includes IL-12p40 and Epstein-Barr virus-induced gene Ebi3. IL-12 members bind to cognate heterodimeric receptor chains expressed on T cells. This family includes IL-12, IL-23, IL-27, IL-35 and IL-39. IL-12 and IL-23 are predominantly proinflammatory cytokines that contribute key roles in the development of Th1 and Th17 cells, respectively. IL-27 has both pro- and anti-inflammatory properties and is a potent T cell immunomodulator.
Catalog No. 50-256-2820 Supplier AdipoGen Supplier No. AG40B0240C050
Recombinant protein. IL-12A/p35 aa 23-215 mouseFc Knobs and IL-12B/p40 aa 23-335 mouseFc Holes form the IL-12 mouseFc-KIH human using the Knobs-into-Holes technology see reference J.B. Ridgway, et al. Protein Eng. 9, 617 1996. Source CHO cells. Lyophilized. Contains PBS Interleukin-12 IL-12 family members are heterodimer glycoproteins, composed of two covalently linked subunits, alpha and beta chains. The alpha-subunit consists of IL-23p19, IL-27p28 and IL-12p35, and the beta-subunit includes IL-12p40 and Epstein-Barr virus-induced gene Ebi3. IL-12 members bind to cognate heterodimeric receptor chains expressed on T cells. This family includes IL-12, IL-23, IL-27, IL-35 and IL-39. IL-12 and IL-23 are predominantly proinflammatory cytokines that contribute key roles in the development of Th1 and Th17 cells, respectively. IL-27 has both pro- and anti-inflammatory properties and is a potent T cell immunomodulator.