Learn More
ACROBiosystems ACROBiosystems Human IL-12B / NKSF2 / p40 Protein, Fc Tag
Supplier:  ACROBiosystems NK2H52581MG
Encompass_Preferred
Recombinant Protein;1MG;Human IL-12B, Fc Tag (NK2-H5258) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Ser 328 (Accession # AAH67499).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 61.3 kDa. The protein migrates as 66-76 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-12B, Fc Tag (Cat. No. NK2-H5258) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-12 R beta 1, Fc,Avitag (Cat. No. ILB-H82F7) with a linear range of 5-40 ng/mL (QC tested).
Catalog No. 50-195-7223