Learn More
ACROBiosystems ACROBiosystems Human IL-12B / NKSF2 / p40 Protein, His Tag
Supplier:  ACROBiosystems NK2H52H71MG
Encompass_Preferred
Recombinant Protein;1MG;Human IL-12B, His Tag (NK2-H52H7) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Ser 328 (Accession # P29460-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.6 kDa. The protein migrates as 44 kDa and 46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti IL23A & IL12B MAb at 2 μg/mL (100 μL/well) can bind Human IL-12B, His Tag (Cat. No. NK2-H52H7) with a linear range of 0.4-13 ng/mL (QC tested).
Catalog No. 50-195-7221