Learn More
ACROBiosystems ACROBiosystems Human IL-12 R beta 1 / CD212 Protein, Fc Tag
Catalog No. 501956854
Encompass_Preferred
Recombinant Protein;100UG;Human IL-12 R beta 1, Fc Tag (ILB-H5255) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 540 (Accession # P42701-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 83.5 kDa. The protein migrates as 90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-12B, His Tag (Cat. No. NK2-H52H7) at 5 μg/mL (100 μL/well) can bind Human IL-12 R beta 1, Fc Tag (Cat. No. ILB-H5255) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 50-195-6854 Supplier ACROBiosystems Supplier No. ILBH5255100UG
Recombinant Protein;100UG;Human IL-12 R beta 1, Fc Tag (ILB-H5255) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 540 (Accession # P42701-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 83.5 kDa. The protein migrates as 90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-12B, His Tag (Cat. No. NK2-H52H7) at 5 μg/mL (100 μL/well) can bind Human IL-12 R beta 1, Fc Tag (Cat. No. ILB-H5255) with a linear range of 5-78 ng/mL (QC tested).