Learn More
ACROBiosystems ACROBiosystems Human IL-12 R beta 1 / CD212 Protein, His Tag
Supplier:  ACROBiosystems ILBH52H91MG
Encompass_Preferred
Recombinant Protein;1MG;Human IL-12 R beta 1, His Tag (ILB-H52H9) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 540 (Accession # P42701-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.0 kDa. The protein migrates as 66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-12 R beta 1, His Tag (Cat. No. ILB-H52H9) at 2 μg/mL (100 μL/well) can bind Human IL-12B, Fc Tag (Cat. No. NK2-H5258) with a linear range of 0.078-1.25 μg/mL (QC tested).
Catalog No. 50-195-7072