Learn More
Medchemexpress LLC HY-P7033 10ug Medchemexpress, IL-13 (CHO-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP703310UG
Encompass_Preferred
Medchemexpress, HY-P7033 10ug IL-13 (CHO-expressed), Human CAS: Recombinant Human Interleukin-13 (CHO-expressed) is a CHO cell derived cytokine which affects the morphology, growth, and surface antigen expression and phenotype of monocytes and elicits B cell proliferation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4280