Learn More
ACROBiosystems ACROBiosystems Human IL-13 Protein, Fc Tag
Catalog No. 501956674
Encompass_Preferred
Recombinant Protein;50UG;Human IL-13, Fc Tag (IL3-H5256) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Asn 132 (Accession # AAK53823.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 38.8 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-13, Fc Tag (Cat. No. IL3-H5256) at 5 μg/mL (100 μL/well) can bind Monoclonal Anti-Human IL-13 Antibody, Human IgG4 with a linear range of 0.4-6 ng/mL (QC tested).;Immobilized Human IL-13, Fc Tag (Cat. No. IL3-H5256) at 5 μg/mL (100 μL/well) can bind Human IL-13 R alpha 2, His Tag (Cat. No. IL2-H52H5) with a linear range of 0.02-0.313 μg/mL (Routinely tested).
Catalog No. 50-195-6674 Supplier ACROBiosystems Supplier No. IL3H525650UG
Recombinant Protein;50UG;Human IL-13, Fc Tag (IL3-H5256) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Asn 132 (Accession # AAK53823.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 38.8 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-13, Fc Tag (Cat. No. IL3-H5256) at 5 μg/mL (100 μL/well) can bind Monoclonal Anti-Human IL-13 Antibody, Human IgG4 with a linear range of 0.4-6 ng/mL (QC tested).;Immobilized Human IL-13, Fc Tag (Cat. No. IL3-H5256) at 5 μg/mL (100 μL/well) can bind Human IL-13 R alpha 2, His Tag (Cat. No. IL2-H52H5) with a linear range of 0.02-0.313 μg/mL (Routinely tested).