Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-13 Protein, His,Avitag™
Catalog No. 501956497
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human IL-13, His,Avitag (IL3-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Asn 132 (Accession # AAK53823.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 15.9 kDa. The protein migrates as 17 kDa and 28-37 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-13 R alpha 2, Fc Tag at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-13, His,Avitag (Cat. No. IL3-H82E5) with a linear range of 0.2-5 ng/mL (QC tested).;Immobilized Biotinylated Human IL-13, His,Avitag (Cat. No. IL3-H82E5) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate can bind Monoclonal Anti-Human IL-13 Antibody, Human IgG4 with a linear range of 0.3-20 ng/mL (Routinely tested).
Catalog No. 50-195-6497 Supplier ACROBiosystems Supplier No. IL3H82E5200UG
Recombinant Protein;200UG;Biotinylated Human IL-13, His,Avitag (IL3-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Asn 132 (Accession # AAK53823.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 15.9 kDa. The protein migrates as 17 kDa and 28-37 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-13 R alpha 2, Fc Tag at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-13, His,Avitag (Cat. No. IL3-H82E5) with a linear range of 0.2-5 ng/mL (QC tested).;Immobilized Biotinylated Human IL-13, His,Avitag (Cat. No. IL3-H82E5) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate can bind Monoclonal Anti-Human IL-13 Antibody, Human IgG4 with a linear range of 0.3-20 ng/mL (Routinely tested).