Learn More
Genscript Corporation Antibody Protein Proteins Proteins 10 UG 1/EA
Supplier:  Genscript Corporation Z0319110
Encompass
Interleukin-13 (IL-13) also known as T-cell activation protein P600 is an immunoregulatory cytokine belonging to the IL-4/IL-13 family. It is produced by activated Th2 cells mast cells and NK cells. IL-13 signals through a receptor complex composed of IL-4RAlpha and IL13RAlpha1 (or IL13RAlpha2). IL-13 inhibits the expression of inflammatory cytokines such as IL-1beta TNF-Alpha and IL-6 by monocytes and macrophages. It also induces B cell activation and IgE secretion.
Catalog No. 50-252-9734