Learn More
Medchemexpress LLC IL-13 Human Biotin 100ug
SDP
Catalog No. 5000181957
Encompass_Preferred
IL-13 protein is a cytokine involved in allergic inflammation immune response to parasites and B cell proliferation IL-13 Protein Human (Biotinylated HEK293 Avi-His) is the recombinant human-derived IL-13 protein expressed by HEK293 with C-Avi C-6 His labeled tag
Catalog No. 50-001-81957 Supplier Medchemexpress LLC Supplier No. HYP72382100UG
IL-13 protein is a cytokine involved in allergic inflammation immune response to parasites and B cell proliferation IL-13 Protein Human (Biotinylated HEK293 Avi-His) is the recombinant human-derived IL-13 protein expressed by HEK293 with C-Avi C-6 His labeled tag