Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-13 ProQuantum Immunoassay Kit

Catalog No. A35609
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A35609 96 Tests
A356095 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A35609 Supplier Invitrogen™ Supplier No. A35609
Add to Cart
Add to Cart

ELISA

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35225
Assay Range 0.0128-5000 pg/mL
Assay Sensitivity <0.01 pg/mL
Content And Storage Contains assay dilution buffer, antibody-conjugate A, antibody-conjugate B, antibody-conjugate dilution buffer, standard, master mix, and ligase. Store at -5°C to -30°C.
Conjugate FAM
Immunoassay Kit Format Proximity-based Immunoassay
Product Type ProQuantum
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) qPCR Instruments
Gene Alias ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600; IL-13; P600
Gene ID (Entrez) 3596
Gene Symbol Il13
Kit Contents Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
Label or Dye FAM™
Protein Aliases IL-13
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL
Shipping Condition Dry Ice
Storage Requirements -20°C
Target IL-13
Test Time 30 min
Total Assay Time 2 hr
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.