Learn More
Enzo Life Sciences IL-13 receptor (human) monoclonal antibody (GM-1C8) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804506C100
Encompass_Preferred
Clone: GM-1C8. Isotype: IgG1. Immunogen: Vector containing the cDNA of the extracellular domain of human IL-13Rα1 (interleukin-13 receptor α1 chain). Source: Purified by affinity chromatography from cell culture supernatant. Species reactivity: Human. Applications: Flow Cytometry. Formulation: Liquid. Antibody in PBS, pH 7.2, containing 0.01% sodium azide.
Catalog No. 50-200-9931