Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human IL-13 R alpha 2 Protein, His Tag
Catalog No. 501956451
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human IL-13 R alpha 2, His Tag (IL2-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Asp 27 - Arg 343 (Accession # Q14627-1). It is the FITC labeled form of Human IL-13 R alpha 2, His Tag (IL2-H52H5).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.0 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-13, His Tag (Cat. No. IL3-H52H4) at 5 μg/mL (100 μL/well) can bind FITC-Labeled Human IL-13 R alpha 2, His Tag (Cat. No. IL2-HF2H3) with a linear range of 0.313-1.25 μg/mL (QC tested).
Catalog No. 50-195-6451 Supplier ACROBiosystems Supplier No. IL2HF2H3200UG
Recombinant Protein;200UG;FITC-Labeled Human IL-13 R alpha 2, His Tag (IL2-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Asp 27 - Arg 343 (Accession # Q14627-1). It is the FITC labeled form of Human IL-13 R alpha 2, His Tag (IL2-H52H5).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.0 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-13, His Tag (Cat. No. IL3-H52H4) at 5 μg/mL (100 μL/well) can bind FITC-Labeled Human IL-13 R alpha 2, His Tag (Cat. No. IL2-HF2H3) with a linear range of 0.313-1.25 μg/mL (QC tested).