Learn More
ACROBiosystems ACROBiosystems Human IL-13 R alpha 2 Protein, His Tag
Supplier:  ACROBiosystems IL2H52H5100UG
Encompass_Preferred
Recombinant Protein;100UG;Human IL-13 R alpha 2, His Tag (IL2-H52H5) is expressed from human 293 cells (HEK293). It contains AA Asp 27 - Arg 343 (Accession # Q14627-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.0 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-13, Fc Tag (Cat. No. IL3-H5256) at 5 μg/mL (100 μL/well) can bind Human IL-13 R alpha 2, His Tag (Cat. No. IL2-H52H5) with a linear range of 4-31 ng/mL (QC tested).
Catalog No. 50-195-6787