Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse IL-15/IL-15R alpha Complex ELISA Kit (Colorimetric)
SDP

Catalog No. NBP192667
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP192667 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP192667 Supplier Novus Biologicals™ Supplier No. NBP192667

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Mouse IL-15/IL-15R alpha Complex ELISA Kit (Colorimetric) measures mouse IL-15/IL-15R alpha Complex in cell culture supernatant, serum, plasma (edta).

Specifications

Assay Range 6.3 - 400pg/mL
Assay Sensitivity 2.0pg/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA), Serum
For Use With (Application) ELISA
Gene ID (Entrez) 3600
Gene Symbol IL15
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-15/IL-15R, Biotin-Conjugate anti-mouse IL-15/IL-15R polyclonal antibody, Streptavidin-HRP, Mouse IL-15/IL-15R Standard lyophilized, 400 pg/ml upon reconstitution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Sample Diluent, Calibrator Diluent, Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym IL-15MGC9721, interleukin 15, interleukin-15
Target IL-15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.