Learn More
ACROBiosystems ACROBiosystems Mouse IL-15 Protein, Fc Tag
Supplier:  ACROBiosystems IL5M52551MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse IL-15, Fc Tag (IL5-M5255) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accession # P48346-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 39.7 kDa. The protein migrates as 49 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse IL-15, Fc Tag (Cat. No. IL5-M5255) at 2 μg/mL (100 μL/well) can bind Biotinylated Human IL-15 R alpha, Fc,Avitag (Cat. No. ILA-H82F4) with a linear range of 10-156 ng/mL (QC tested).
Catalog No. 50-195-6806