Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-15 Protein, Fc,Avitag™
Supplier:  ACROBiosystems IL5H82F325UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human IL-15, Fc,Avitag (IL5-H82F3) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accession # P40933-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 41.0 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-15 R alpha, Fc Tag (Cat. No. ILA-H5253) at 2 μg/mL (100 μL/well) can bind Biotinylated Human IL-15, Fc,Avitag (Cat. No. IL5-H82F3) with a linear range of 2-78 ng/mL (QC tested).
Catalog No. 50-195-7038