Learn More
ACROBiosystems ACROBiosystems Human IL-15 Protein, His Tag
Supplier:  ACROBiosystems IL5H52H850UG
Encompass_Preferred
Recombinant Protein;50UG;Human IL-15, His Tag (IL5-H52H8) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accession # P40933-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.7 kDa.
Catalog No. 50-195-6382