Learn More
ACROBiosystems ACROBiosystems Human IL-15 Protein, His Tag
Catalog No. 501956381
Encompass_Preferred
Recombinant Protein;1MG;Human IL-15, His Tag (IL5-H52H8) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accession # P40933-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.7 kDa.
Catalog No. 50-195-6381 Supplier ACROBiosystems Supplier No. IL5H52H81MG
Recombinant Protein;1MG;Human IL-15, His Tag (IL5-H52H8) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accession # P40933-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.7 kDa.