Learn More
Enzo Life Sciences IL-15 (human), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2550100
Encompass_Preferred
Alternative name: Interleukin-15, IL-T. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium bicarbonate, pH 8. Source: Produced in E. coli. Contains 115 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4071