Learn More
Sigma Aldrich Fine Chemicals Biosciences IL 15 human Animal compone
Catalog No. 502905079
Encompass
Interleukin-15 (IL-15) is an immunomodulating cytokine. The gene encoding it is localized on chromosome 4q31. Human IL-15 is biologically active on mouse cells as measured by the dose-dependent stimulation of the proliferation of mouse CTLL cells. Recombinant human IL-15 is a 12.9kDa protein consisting of 115 amino acid residues.
Catalog No. 50-290-5079 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRP307710UG
Interleukin-15 (IL-15) is an immunomodulating cytokine. The gene encoding it is localized on chromosome 4q31. Human IL-15 is biologically active on mouse cells as measured by the dose-dependent stimulation of the proliferation of mouse CTLL cells. Recombinant human IL-15 is a 12.9kDa protein consisting of 115 amino acid residues.