Learn More
Medchemexpress LLC IL-15 Human Biotin 50ug | 50ug
SDP
Catalog No. 5000194893
Encompass_Preferred
IL-15 protein regulates immune cells stimulates the proliferation of NK cells T cells and B cells and promotes cytokine secretion IL-15 Protein Human (Biotinylated HEK293 Fc-Avi) is the recombinant human-derived IL-15 protein expressed by HEK293 with C-Avi C-hFc labeled tag
Catalog No. 50-001-94893 Supplier Medchemexpress LLC Supplier No. HYP7886050UG
IL-15 protein regulates immune cells stimulates the proliferation of NK cells T cells and B cells and promotes cytokine secretion IL-15 Protein Human (Biotinylated HEK293 Fc-Avi) is the recombinant human-derived IL-15 protein expressed by HEK293 with C-Avi C-hFc labeled tag