Learn More
Medchemexpress LLC IL-15 Human HEK293 20ug
SDP
Catalog No. 5000185492
Encompass_Preferred
Recombinant human interleukin-15 (IL-15) produced in HEK293 cells and supplied as a lyophilized, tag-free protein for research use. The preparation corresponds to residues N49-S162 of the human IL-15 sequence (accession P40933-1), is glycosylated, and is characterized for purity, endotoxin, and biological activity to support in vitro and cell culture assays. Store lyophilized at -20°C and follow recommended reconstitution and aliquoting procedures.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • Tag free and matches the native sequence (N49-S162).
  • High purity (95% by reducing SDS-PAGE).
  • Low endotoxin level (<1 EU/μg) suitable for cell-based assays.
  • Verified biological activity (0.95 x 10^7 IU/mg).
  • Lyophilized in PBS with 11% trehalose for enhanced stability.
  • Stable frozen at -20°C; reconstitute at ≥100 μg/mL and aliquot for long-term storage.
Catalog No. 50-001-85492 Supplier Medchemexpress LLC Supplier No. HYP70469220UG
Recombinant human interleukin-15 (IL-15) produced in HEK293 cells and supplied as a lyophilized, tag-free protein for research use. The preparation corresponds to residues N49-S162 of the human IL-15 sequence (accession P40933-1), is glycosylated, and is characterized for purity, endotoxin, and biological activity to support in vitro and cell culture assays. Store lyophilized at -20°C and follow recommended reconstitution and aliquoting procedures.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • Tag free and matches the native sequence (N49-S162).
  • High purity (95% by reducing SDS-PAGE).
  • Low endotoxin level (<1 EU/μg) suitable for cell-based assays.
  • Verified biological activity (0.95 x 10^7 IU/mg).
  • Lyophilized in PBS with 11% trehalose for enhanced stability.
  • Stable frozen at -20°C; reconstitute at ≥100 μg/mL and aliquot for long-term storage.