Learn More
Enzo Life Sciences IL-15 (mouse), (recombinant) (10µg)
SDP
Catalog No. 502013931
Encompass_Preferred
Alternative name: Interleukin-15. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized from 10mM sodium phosphate, pH 8.0. Source: Produced in E.coli. Long Term Storage: -20°C.
Catalog No. 50-201-3931 Supplier Enzo Life Sciences Supplier No. ENZPRT1400010
Alternative name: Interleukin-15. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized from 10mM sodium phosphate, pH 8.0. Source: Produced in E.coli. Long Term Storage: -20°C.