Learn More
ACROBiosystems ACROBiosystems Human IL-15 R alpha / CD215 Protein, Fc Tag
Catalog No. 501956940
Encompass_Preferred
Recombinant Protein;1MG;Human IL-15 R alpha, Fc Tag (ILA-H5253) is expressed from human 293 cells (HEK293). It contains AA Ile 31 - Thr 205 (Accession # Q13261-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 44.8 kDa. The protein migrates as 60-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-15 R alpha, Fc Tag (Cat. No. ILA-H5253) at 2 μg/mL (100 μL/well) can bind Biotinylated Human IL-15, Fc,Avitag (Cat. No. IL5-H82F3) with a linear range of 2-78 ng/mL (QC tested).;Human IL-15 R alpha, Fc Tag (Cat. No. ILA-H5253) inhibits IL-15-dependent proliferation of CTLL-2 cells. The ED50 for this effect is 2.77-4.20 ng/mL (Routinely tested).
Catalog No. 50-195-6940 Supplier ACROBiosystems Supplier No. ILAH52531MG
Recombinant Protein;1MG;Human IL-15 R alpha, Fc Tag (ILA-H5253) is expressed from human 293 cells (HEK293). It contains AA Ile 31 - Thr 205 (Accession # Q13261-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 44.8 kDa. The protein migrates as 60-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-15 R alpha, Fc Tag (Cat. No. ILA-H5253) at 2 μg/mL (100 μL/well) can bind Biotinylated Human IL-15, Fc,Avitag (Cat. No. IL5-H82F3) with a linear range of 2-78 ng/mL (QC tested).;Human IL-15 R alpha, Fc Tag (Cat. No. ILA-H5253) inhibits IL-15-dependent proliferation of CTLL-2 cells. The ED50 for this effect is 2.77-4.20 ng/mL (Routinely tested).