Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-15 R alpha / CD215 Protein, Fc,Avitag™
Supplier:  ACROBiosystems ILAH82F4200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human IL-15 R alpha, Fc,Avitag (ILA-H82F4) is expressed from human 293 cells (HEK293). It contains AA Ile 31 - Thr 205 (Accession # Q13261-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 46.6 kDa. The protein migrates as 60-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-15, Tag Free (Cat. No. IL5-H4117) at 2 μg/mL (100 μL/well) can bind Biotinylated Human IL-15 R alpha, Fc,Avitag (Cat. No. ILA-H82F4) with a linear range of 2-39 ng/mL (QC tested).;Immobilized Mouse IL-15, Fc Tag (Cat. No. IL5-M5255) at 2 μg/mL (100 μL/well) can bind Biotinylated Human IL-15 R alpha, Fc,Avitag (Cat. No. ILA-H82F4) with a linear range of 10-156 ng/mL (Routinely tested).
Catalog No. 50-195-6900