Learn More
Medchemexpress LLC IL-17 modulator 1 disodium | 2446803-91-2 | 98.4% | 628.57 | C28H35N6Na2O6P | 5 MG
SDP
Supplier:  Medchemexpress LLC HY141535A5MG
Encompass_Preferred
IL-17 modulator 1 (disodium) is an orally active small-molecule modulator of interleukin-17 intended for preclinical research. It is described for use in studies of IL-17-mediated diseases such as psoriasis, ankylosing spondylitis, and psoriatic arthritis, and is supplied as a solid powder or as a 10 mM solution in DMSO. Intended for research use only.
  • Orally active small-molecule IL-17 modulator.
  • Disodium salt form for improved solubility.
  • Available as solid powder and 10 mM solution in DMSO.
  • High purity (~98.4%) suitable for analytical work.
  • Suitable for in vitro and in vivo preclinical studies.
  • Documented molecular weight and formula for characterization.
Catalog No. 50-002-23177