Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-17 modulator 1 disodium | 2446803-91-2 | 98.4% | C28H35N6Na2O6P | 10MG
SDP

Catalog No. 5000261746
Encompass_Preferred

IL-17 modulator 1 disodium is an orally active small-molecule research compound that modulates interleukin-17 (IL-17) signaling and is used in preclinical studies of IL-17-mediated inflammatory diseases such as psoriasis, psoriatic arthritis, and ankylosing spondylitis. Supplied as a disodium salt powder with documented chemical identity and analytical data for research use only.

  • Orally active small-molecule IL-17 pathway modulator.
  • Disodium salt form suitable for formulation studies.
  • Chemical formula C28H35N6Na2O6P and molecular weight 628.57.
  • High purity (~98.4%) as provided by supplier.
  • Available in small research quantities for preclinical use.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-61746 Supplier Medchemexpress LLC Supplier No. HY141535A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IL-17 modulator 1 disodium is an orally active small-molecule research compound that modulates interleukin-17 (IL-17) signaling and is used in preclinical studies of IL-17-mediated inflammatory diseases such as psoriasis, psoriatic arthritis, and ankylosing spondylitis. Supplied as a disodium salt powder with documented chemical identity and analytical data for research use only.

  • Orally active small-molecule IL-17 pathway modulator.
  • Disodium salt form suitable for formulation studies.
  • Chemical formula C28H35N6Na2O6P and molecular weight 628.57.
  • High purity (~98.4%) as provided by supplier.
  • Available in small research quantities for preclinical use.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.