Learn More
Medchemexpress LLC IL-17 modulator 1 (disodium) | 2446803-91-2 | 98.4% | C28H35N6Na2O6P | 25 MG
SDP
Catalog No. 5000357288
Encompass_Preferred
IL-17 modulator 1 disodium is an orally active, highly efficacious small molecule IL-17 modulator. It is designed for research into diseases such as psoriasis, ankylosing spondylitis, and psoriatic arthritis. The disodium salt form provides enhanced water solubility and stability, making it suitable for various research applications.
  • Orally active and highly efficacious small molecule
  • Enhanced water solubility and stability
  • Useful for research into psoriasis
  • Useful for research into ankylosing spondylitis
  • Useful for research into psoriatic arthritis
Catalog No. 50-003-57288 Supplier Medchemexpress LLC Supplier No. 25MGHY141535A
IL-17 modulator 1 disodium is an orally active, highly efficacious small molecule IL-17 modulator. It is designed for research into diseases such as psoriasis, ankylosing spondylitis, and psoriatic arthritis. The disodium salt form provides enhanced water solubility and stability, making it suitable for various research applications.
  • Orally active and highly efficacious small molecule
  • Enhanced water solubility and stability
  • Useful for research into psoriasis
  • Useful for research into ankylosing spondylitis
  • Useful for research into psoriatic arthritis