Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-17 modulator 4 | 2446803-65-0 | 99.0% | 474.6 g/mol | C27H34N6O2 | 5 MG
SDP

Catalog No. 5000221181
Encompass_Preferred

IL-17 modulator 4 is a small-molecule proagent for research of IL-17A-mediated diseases, including inflammation, autoimmune disorders, infectious disease, and cancer. Available as a solid or as a DMSO solution for in vitro studies, it is supplied with analytical purity and documented storage and handling recommendations.

  • High purity (99.0%).
  • Molecular weight 474.6 g/mol.
  • Chemical formula C27H34N6O2.
  • Soluble in DMSO (≈300 mg/mL); ultrasonic agitation recommended.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years).
  • In solvent: store at -80°C for up to 6 months or -20°C for up to 1 month.
  • Supplied in multiple pack sizes and solution formats for assay flexibility.

Catalog No. 50-002-21181 Supplier Medchemexpress LLC Supplier No. HY1416925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IL-17 modulator 4 is a small-molecule proagent for research of IL-17A-mediated diseases, including inflammation, autoimmune disorders, infectious disease, and cancer. Available as a solid or as a DMSO solution for in vitro studies, it is supplied with analytical purity and documented storage and handling recommendations.

  • High purity (99.0%).
  • Molecular weight 474.6 g/mol.
  • Chemical formula C27H34N6O2.
  • Soluble in DMSO (≈300 mg/mL); ultrasonic agitation recommended.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years).
  • In solvent: store at -80°C for up to 6 months or -20°C for up to 1 month.
  • Supplied in multiple pack sizes and solution formats for assay flexibility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.