Learn More
Medchemexpress LLC IL-17 modulator 4 | 2446803-65-0 | >99.0% | C27H34N6O2 | 1 MG
SDP
Catalog No. 5000353406
Encompass_Preferred
IL-17 modulator 4 is a proagent of IL-17 modulator 1, an orally active and highly efficacious IL-17 modulator. This compound is promising for the research of IL-17A mediated diseases, including inflammation, autoimmune diseases, infectious diseases, cancer, and precancerous syndrome. It modulates IL-17A protein-protein interaction by stabilizing the conformation of the IL-17A dimer, preventing binding to the IL-17RA receptor and inhibiting the IL-17A-mediated signaling pathway. It demonstrates favorable pharmacokinetic properties with excellent oral bioavailability.
  • Modulates IL-17A protein-protein interaction
  • Stabilizes IL-17A dimer conformation
  • Inhibits IL-17A-mediated signaling pathway
  • Potential for research in inflammation and autoimmune diseases
  • Exhibits good pharmacokinetic properties in rat models
  • Offers high oral bioavailability
Catalog No. 50-003-53406 Supplier Medchemexpress LLC Supplier No. HY1416921MG
IL-17 modulator 4 is a proagent of IL-17 modulator 1, an orally active and highly efficacious IL-17 modulator. This compound is promising for the research of IL-17A mediated diseases, including inflammation, autoimmune diseases, infectious diseases, cancer, and precancerous syndrome. It modulates IL-17A protein-protein interaction by stabilizing the conformation of the IL-17A dimer, preventing binding to the IL-17RA receptor and inhibiting the IL-17A-mediated signaling pathway. It demonstrates favorable pharmacokinetic properties with excellent oral bioavailability.
  • Modulates IL-17A protein-protein interaction
  • Stabilizes IL-17A dimer conformation
  • Inhibits IL-17A-mediated signaling pathway
  • Potential for research in inflammation and autoimmune diseases
  • Exhibits good pharmacokinetic properties in rat models
  • Offers high oral bioavailability