Learn More
Medchemexpress LLC IL-17 modulator 4 | 2446803-65-0 | >99.0% | C27H34N6O2 | 1 ML
SDP
Catalog No. 5000353408
Encompass_Preferred
IL-17 modulator 4 is a proagent of IL-17 modulator 1, active against IL-17A mediated diseases. This orally active modulator shows promise for the research of conditions including inflammation, autoimmune diseases, infectious diseases, cancer, and precancerous syndrome. It inhibits IL-17A-mediated signaling pathways by stabilizing the IL-17A dimer and preventing its binding to the IL-17RA receptor.
  • Modulates IL-17A protein-protein interaction
  • Stabilizes IL-17A dimer conformation
  • Prevents IL-17A from binding to IL-17RA receptor
  • Inhibits IL-17A-mediated signaling pathways
  • Exhibits good pharmacokinetic properties in rat models
  • Oral bioavailability of 100% in rat models
Catalog No. 50-003-53408 Supplier Medchemexpress LLC Supplier No. HY14169210MM/1ML
IL-17 modulator 4 is a proagent of IL-17 modulator 1, active against IL-17A mediated diseases. This orally active modulator shows promise for the research of conditions including inflammation, autoimmune diseases, infectious diseases, cancer, and precancerous syndrome. It inhibits IL-17A-mediated signaling pathways by stabilizing the IL-17A dimer and preventing its binding to the IL-17RA receptor.
  • Modulates IL-17A protein-protein interaction
  • Stabilizes IL-17A dimer conformation
  • Prevents IL-17A from binding to IL-17RA receptor
  • Inhibits IL-17A-mediated signaling pathways
  • Exhibits good pharmacokinetic properties in rat models
  • Oral bioavailability of 100% in rat models