Learn More
Medchemexpress LLC IL-17 modulator 4 | 2446803-65-0 | 99.0% | C27H34N6O2 | 25 MG
SDP
Catalog No. 5000357404
Encompass_Preferred
IL-17 modulator 4 is a proagent of IL-17 modulator 1, an orally active and highly efficacious IL-17 modulator, promising for research into IL-17A mediated diseases like inflammation, autoimmune diseases, infectious diseases, cancer, and precancerous syndrome.
  • Proagent of an orally active, highly efficacious IL-17 modulator.
  • Promising for research into IL-17A mediated diseases.
  • Modulates IL-17A protein-protein interaction by stabilizing its conformation and preventing binding to the IL-17RA receptor.
  • Inhibits the IL-17A-mediated signaling pathway and IL-8 secretion in HEKa cells (EC50 of 14 nM).
  • Exhibits good pharmacokinetic properties in rat models (low clearance, moderate volume of distribution, 3.7 h half-life, 100% oral bioavailability).
  • Appearance: solid, off-white to gray.
Catalog No. 50-003-57404 Supplier Medchemexpress LLC Supplier No. 25MGHY141692
IL-17 modulator 4 is a proagent of IL-17 modulator 1, an orally active and highly efficacious IL-17 modulator, promising for research into IL-17A mediated diseases like inflammation, autoimmune diseases, infectious diseases, cancer, and precancerous syndrome.
  • Proagent of an orally active, highly efficacious IL-17 modulator.
  • Promising for research into IL-17A mediated diseases.
  • Modulates IL-17A protein-protein interaction by stabilizing its conformation and preventing binding to the IL-17RA receptor.
  • Inhibits the IL-17A-mediated signaling pathway and IL-8 secretion in HEKa cells (EC50 of 14 nM).
  • Exhibits good pharmacokinetic properties in rat models (low clearance, moderate volume of distribution, 3.7 h half-life, 100% oral bioavailability).
  • Appearance: solid, off-white to gray.