Learn More
Medchemexpress LLC HY-P7372 10ug Medchemexpress, IL-17A (CHO-expressed), Mouse CAS: Purity:>98%
SDP
Catalog No. 501934384
Encompass_Preferred
Medchemexpress, HY-P7372 10ug IL-17A (CHO-expressed), Mouse CAS: Recombinant Mouse Interleukin-17A (CHO-expressed) is a cytokine that plays an important role in both innate and acquired immunity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4384 Supplier Medchemexpress LLC Supplier No. HYP737210UG
Medchemexpress, HY-P7372 10ug IL-17A (CHO-expressed), Mouse CAS: Recombinant Mouse Interleukin-17A (CHO-expressed) is a cytokine that plays an important role in both innate and acquired immunity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.