Learn More
ACROBiosystems ACROBiosystems Mouse IL-17A / CTLA-8 Protein, His Tag
Catalog No. 501957279
Encompass_Preferred
Recombinant Protein;1MG;Mouse IL-17A, His Tag (CT8-M5240) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Ala 158 (Accession # Q62386-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 15.8 kDa. The protein migrates as 18 kDa, 20-23 kDa and 26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse IL-17A, His Tag (Cat. No. CT8-M5240) at 2 μg/mL (100 μL/well) can bind Human IL-17 RA, Fc Tag (Cat. No. ILA-H5257) with a linear range of 8-128 ng/mL (QC tested).;Mouse IL-17A, His Tag (Cat. No. CT8-M5240) stimulates secretion of IL-6 by NIH-3T3 cells. The ED50 for this effect is 2.132-5.005 ng/mL.
Catalog No. 50-195-7279 Supplier ACROBiosystems Supplier No. CT8M52401MG
Recombinant Protein;1MG;Mouse IL-17A, His Tag (CT8-M5240) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Ala 158 (Accession # Q62386-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 15.8 kDa. The protein migrates as 18 kDa, 20-23 kDa and 26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse IL-17A, His Tag (Cat. No. CT8-M5240) at 2 μg/mL (100 μL/well) can bind Human IL-17 RA, Fc Tag (Cat. No. ILA-H5257) with a linear range of 8-128 ng/mL (QC tested).;Mouse IL-17A, His Tag (Cat. No. CT8-M5240) stimulates secretion of IL-6 by NIH-3T3 cells. The ED50 for this effect is 2.132-5.005 ng/mL.