Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-17A / CTLA-8 Protein, His,Avitag™
Supplier:  ACROBiosystems ILAH82Q1200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human IL17A, His,Avitag (ILA-H82Q1) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (Accession # Q16552-1).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 17.8 kDa. The protein migrates as 19-21 kDa and 23-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-17 RA, Fc Tag (Cat. No. ILA-H5257) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL17A, His,Avitag (Cat. No. ILA-H82Q1) with a linear range of 0.3-10 ng/mL (QC tested).;Immobilized Monoclonal Anti-Human IL-17A Antibody, Human IgG1 at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL17A, His,Avitag (Cat. No. ILA-H82Q1) with a linear range of 0.3-2 ng/mL (Routinely tested).
Catalog No. 50-195-6718