Learn More
Medchemexpress LLC IL-17A Human CHO 100ug
SDP
Catalog No. 5000182271
Encompass_Preferred
IL-17A is a homodimeric cytokine and plays a critical role in host defense mechanisms against many bacterial and fungal pathogens as well as allergic and autoimmune responses IL-17A induces the production of antimicrobial peptides cytokines chemokines and matrix metalloproteinases[1][2] IL-17A Protein Human (CHO) is a recombinant human IL-17A protein and is expressed in CHO cells It consists of 155 amino acids (M1-A155)
Catalog No. 50-001-82271 Supplier Medchemexpress LLC Supplier No. HYP73171100UG
IL-17A is a homodimeric cytokine and plays a critical role in host defense mechanisms against many bacterial and fungal pathogens as well as allergic and autoimmune responses IL-17A induces the production of antimicrobial peptides cytokines chemokines and matrix metalloproteinases[1][2] IL-17A Protein Human (CHO) is a recombinant human IL-17A protein and is expressed in CHO cells It consists of 155 amino acids (M1-A155)