Learn More
TARGETMOL CHEMICALS INC IL-17A inhibitor 1 1MG
SDP
Catalog No. 502877260
Encompass_Preferred
Also available in 1 mL and bulk. Please contact Fisher for quotes. IL-17A inhibitor 1 is a IL-17A inhibitor with IC50 of <9.45 nM in alphalisa assay. IL-17A inhibitor 1 inhibits HT-29 cells with IC50 of 9.3 nM. Purity 99.38%
Catalog No. 50-287-7260 Supplier TARGETMOL CHEMICALS INC Supplier No. T94481MG
Also available in 1 mL and bulk. Please contact Fisher for quotes. IL-17A inhibitor 1 is a IL-17A inhibitor with IC50 of <9.45 nM in alphalisa assay. IL-17A inhibitor 1 inhibits HT-29 cells with IC50 of 9.3 nM. Purity 99.38%