Learn More
Medchemexpress LLC IL-17A modulator-1 | 2748749-29-1 | 98.8% | C33H31N5O4 | 10MG
SDP
Catalog No. 5000261469
Encompass_Preferred
IL-17A modulator-1 is a small-molecule research compound that inhibits IL-17A biological activity (reported pIC50 = 8.2). Provided as a solid with high analytical purity, it is intended for use as a tool compound in studies of IL-17A-related biology and disease models.
  • Reported activity: pIC50 = 8.2 against IL-17A.
  • Molecular formula C33H31N5O4 and molecular weight 561.63.
  • High purity (98.8%) suitable for research applications.
  • Available in multiple pack sizes to support assay scale-up.
  • Solid form for convenient handling and formulation.
Catalog No. 50-002-61469 Supplier Medchemexpress LLC Supplier No. HY14543010MG
IL-17A modulator-1 is a small-molecule research compound that inhibits IL-17A biological activity (reported pIC50 = 8.2). Provided as a solid with high analytical purity, it is intended for use as a tool compound in studies of IL-17A-related biology and disease models.
  • Reported activity: pIC50 = 8.2 against IL-17A.
  • Molecular formula C33H31N5O4 and molecular weight 561.63.
  • High purity (98.8%) suitable for research applications.
  • Available in multiple pack sizes to support assay scale-up.
  • Solid form for convenient handling and formulation.