Learn More
Medchemexpress LLC IL-17A modulator-1 | 2748749-29-1 | 98.8% | C33H31N5O4 | 25 MG
SDP
Catalog No. 5000353245
Encompass_Preferred
IL-17A modulator-1 is a compound that modulates the activity of IL-17A, an important cytokine involved in immune responses. Derived from patent WO2021239743+A1, this modulator inhibits the biological action of IL-17A with a pIC50 of 8.2, making it suitable for research into various diseases and disorders associated with IL-17A activity, including those with immune components, autoimmune pathologies, cancer, and neurodegenerative disorders. The product is provided as a white to off-white solid.
  • Modulates IL-17A activity
  • Inhibits biological action of IL-17A
  • Suitable for research into immune, autoimmune, cancer, and neurodegenerative disorders
  • White to off-white solid appearance
Catalog No. 50-003-53245 Supplier Medchemexpress LLC Supplier No. 25MGHY145430
IL-17A modulator-1 is a compound that modulates the activity of IL-17A, an important cytokine involved in immune responses. Derived from patent WO2021239743+A1, this modulator inhibits the biological action of IL-17A with a pIC50 of 8.2, making it suitable for research into various diseases and disorders associated with IL-17A activity, including those with immune components, autoimmune pathologies, cancer, and neurodegenerative disorders. The product is provided as a white to off-white solid.
  • Modulates IL-17A activity
  • Inhibits biological action of IL-17A
  • Suitable for research into immune, autoimmune, cancer, and neurodegenerative disorders
  • White to off-white solid appearance